2-Bromo-5-fluorobenzyl fluoride 95%
Proveedor: Apollo Scientific
|
Peligro
|
PC27928-1G
PC27928-250MG
APOSPC27928-1GEA
671
EUR
APOSPC27928-1G
APOSPC27928-250MG
2-Bromo-5-fluorobenzyl fluoride 95%
2-Bromo-5-fluorobenzyl fluoride
Formula:
C₇H₅BrF₂ MW: 207,015 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD31664907 Núm. CAS: 2244085-04-7 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...