3-Butoxy-2,6-difluorophenylacetic acid 97%
Proveedor: Apollo Scientific
PC303119-1G
PC303119-5G
APOSPC303119-1GEA
226
EUR
APOSPC303119-1G
APOSPC303119-5G
3-Butoxy-2,6-difluorophenylacetic acid 97%
3-Butoxy-2,6-difluorophenylacetic acid
Formula:
C₁₂H₁₄F₂O₃ MW: 244,235 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD27986844 Núm. CAS: 1706446-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...