3,4-Dichloro-5-(trifluoromethoxy)benzoyl chloride 97%
Proveedor: Apollo Scientific
PC303171-1G
PC303171-250MG
APOSPC303171-1GEA
620
EUR
APOSPC303171-1G
APOSPC303171-250MG
3,4-Dichloro-5-(trifluoromethoxy)benzoyl chloride 97%
3,4-Dichloro-5-(trifluoromethoxy)benzoyl chloride
Formula:
C₈H₂Cl₃F₃O₂ MW: 293,454 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD28054195 Núm. CAS: 1706458-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...