4-Chloro-2-fluoro-5-methylbenzoic acid 97%
Proveedor: Apollo Scientific
PC303325-1G
PC303325-250MG
APOSPC303325-1GEA
299
EUR
APOSPC303325-1G
APOSPC303325-250MG
4-Chloro-2-fluoro-5-methylbenzoic acid 97%
4-Chloro-2-fluoro-5-methylbenzoic acid
Formula:
C₈H₆ClFO₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD20660427 Núm. CAS: 177211-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...