Ethyl 3-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl]propionate
Proveedor: Apollo Scientific
PC401028-1G
PC401028-5G
APOSPC401028-1GEA
259
EUR
APOSPC401028-1G
APOSPC401028-5G
Ethyl 3-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl]propionate
Ethyl 3-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]piperazin-1-yl]propionate
Formula:
C₁₅H₁₉ClF₃N₃O₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD28122851 Núm. CAS: 1672655-81-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...