3-Fluoro-4-(4-formylphenoxy)benzonitrile
Proveedor: Apollo Scientific
PC402139-1G
PC402139-250MG
APOSPC402139-1GEA
560
EUR
APOSPC402139-1G
APOSPC402139-250MG
3-Fluoro-4-(4-formylphenoxy)benzonitrile
3-Fluoro-4-(4-formylphenoxy)benzonitrile
Formula:
C₁₄H₈FNO₂ MW: 241,217 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD11111105 Núm. CAS: 676494-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...