3-Fluorobenzoyl isocyanate
Proveedor: Apollo Scientific
PC404018-1G
PC404018-250MG
APOSPC404018-1GEA
509
EUR
APOSPC404018-1G
APOSPC404018-250MG
3-Fluorobenzoyl isocyanate
3-Fluorobenzoyl isocyanate
May form small amount of sediment
Formula:
C₈H₄FNO₂ MW: 165,121 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD09816474 Núm. CAS: 78469-21-3 UN: 2206 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...