1-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]methanamine hydrochloride

Proveedor: Apollo Scientific

PC408452-1G PC408452-250MG PC408452-5G
APOSPC408452-1GEA 712 EUR
APOSPC408452-1G APOSPC408452-250MG APOSPC408452-5G
1-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]methanamine hydrochloride
1-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]methanamine hydrochloride
Formula: C₆H₈F₃N₃·ClH
Temperatura de almacenaje: Ambiente
Número MDL: MFCD28893666
Núm. CAS: 2138274-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR