Methyl 5-fluoro-1H-indole-2-carboxylate 95+%

Proveedor: Apollo Scientific

PC410179-1G PC410179-5G
APOSPC410179-1GEA 412 EUR
APOSPC410179-1G APOSPC410179-5G
Methyl 5-fluoro-1H-indole-2-carboxylate 95+%
Methyl 5-fluoro-1H-indole-2-carboxylate
Formula: C₁₀H₈FNO₂
MW: 193,174 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD04972072
Núm. CAS: 167631-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR