3-(Fluoromethyl)azetidine hydrochloride
Proveedor: Apollo Scientific
PC411261-1G
PC411261-250MG
APOSPC411261-1GEA
509
EUR
APOSPC411261-1G
APOSPC411261-250MG
3-(Fluoromethyl)azetidine hydrochloride
3-(Fluoromethyl)azetidine hydrochloride
Formula:
C₄H₈FN·ClH MW: 125,572 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD27991715 Núm. CAS: 1642298-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...