2-Chloro-4,6-difluorobenzene boronic acid 95%
Proveedor: Apollo Scientific
Sinónimos:
(2-Chloro-4,6-difluorophenyl)boronic acid
PC44901-1G
PC44901-250MG
APOSPC44901-1GEA
522
EUR
APOSPC44901-1G
APOSPC44901-250MG
2-Chloro-4,6-difluorobenzene boronic acid 95%
2-Chloro-4,6-difluorobenzene boronic acid
Formula:
C₆H₄BClF₂O₂ MW: 192,356 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD18415645 Núm. CAS: 1373393-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...