Bis(nonafluorobutylsulphonyl)methane
Proveedor: Apollo Scientific
PC450104-1G
PC450104-250MG
APOSPC450104-1GEA
723
EUR
APOSPC450104-1G
APOSPC450104-250MG
Bis(nonafluorobutylsulphonyl)methane
Bis(nonafluorobutylsulphonyl)methane
Formula:
C₉H₂F₁₈O₄S₂ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD20489412 Núm. CAS: 29214-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...