2-Fluoro-6-(trifluoromethyl)benzylamine hydrochloride

Proveedor: Apollo Scientific

PC450151-25G PC450151-5G PC450151-1G
APOSPC450151-25GEA 336 EUR
APOSPC450151-25G APOSPC450151-5G APOSPC450151-1G
2-Fluoro-6-(trifluoromethyl)benzylamine hydrochloride
2-Fluoro-6-(trifluoromethyl)benzylamine hydrochloride
Formula: C₈H₇F₄N·ClH
Número MDL: MFCD08437622
Núm. CAS: 643088-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR