Difluoro(pyridin-2-yl)acetic acid hydrochloride
Proveedor: Apollo Scientific
|
Atención
|
Sinónimos:
2-[Carboxy(difluoro)methyl]pyridine hydrochloride, 2,2-Difluoro-2-(pyridin-2-yl)ethanoic acid hydrochloride
PC49343-1G
PC49343-5G
APOSPC49343-1GEA
140
EUR
APOSPC49343-1G
APOSPC49343-5G
Difluoro(pyridin-2-yl)acetic acid hydrochloride
Difluoro(pyridin-2-yl)acetic acid hydrochloride
Formula:
C₇H₅F₂NO₂·ClH MW: 209,578 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD22392210 Núm. CAS: 1421603-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...