3-Bromo-6-fluoro-2-nitrobenzyl bromide
Proveedor: Apollo Scientific
PC500371-1G
PC500371-5G
APOSPC500371-1GEA
437
EUR
APOSPC500371-1G
APOSPC500371-5G
3-Bromo-6-fluoro-2-nitrobenzyl bromide
3-Bromo-6-fluoro-2-nitrobenzyl bromide
Formula:
C₇H₄Br₂FNO₂ MW: 312,919 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD27975445 Núm. CAS: 1807263-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...