4-Bromo-2-iodobenzyl fluoride 95%
Proveedor: Apollo Scientific
|
Peligro
|
PC502889-1G
PC502889-250MG
APOSPC502889-1GEA
609
EUR
APOSPC502889-1G
APOSPC502889-250MG
4-Bromo-2-iodobenzyl fluoride 95%
4-Bromo-2-iodobenzyl fluoride
Formula:
C₇H₅BrFI MW: 314,921 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD22491802 Núm. CAS: 1823057-30-2 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...