3-Bromo-2-fluorophenylisocyanate

Proveedor: Apollo Scientific
Peligro

PC503006-1G PC503006-25G PC503006-5G
APOSPC503006-1GEA 21.9 EUR
APOSPC503006-1G APOSPC503006-25G APOSPC503006-5G
3-Bromo-2-fluorophenylisocyanate
3-Bromo-2-fluorophenylisocyanate

May form small amount of sediment

Formula: C₇H₃BrFNO
Temperatura de almacenaje: Nevera
Número MDL: MFCD26679158
Núm. CAS: 1506860-05-4
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR