Ethyl 3-(2,2,2-Trifluoro-1-hydroxyethyl)benzoate
Proveedor: Apollo Scientific
PC510280-1G
PC510280-5G
APOSPC510280-1GEA
396
EUR
APOSPC510280-1G
APOSPC510280-5G
Ethyl 3-(2,2,2-Trifluoro-1-hydroxyethyl)benzoate
Ethyl 3-(2,2,2-Trifluoro-1-hydroxyethyl)benzoate
Formula:
C₁₁H₁₁F₃O₃ MW: 248,198 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD29035075 Núm. CAS: 2006277-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...