4-(Aminomethyl)-2-(trifluoromethyl)aniline hydrochloride
Proveedor: Apollo Scientific
PC510295-1G
PC510295-5G
APOSPC510295-1GEA
205
EUR
APOSPC510295-1G
APOSPC510295-5G
4-(Aminomethyl)-2-(trifluoromethyl)aniline hydrochloride
4-(Aminomethyl)-2-(trifluoromethyl)aniline hydrochloride
Formula:
C₈H₉F₃N₂·ClH MW: 226,627 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD28122820 Núm. CAS: 1196702-79-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...