1-Fluoro-2-propylamine hydrochloride 97%
Proveedor: Apollo Scientific
Sinónimos:
1-Fluoropropan-2-amine hydrochloride
PC510357-100MG
PC510357-250MG
APOSPC510357-100MGEA
116
EUR
APOSPC510357-100MG
APOSPC510357-250MG
1-Fluoro-2-propylamine hydrochloride 97%
1-Fluoro-2-propylamine hydrochloride
Formula:
C₃H₈FN·ClH MW: 113,562 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD20489620 Núm. CAS: 921602-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...