Methyl 4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononanoate
Proveedor: Apollo Scientific
Sinónimos:
Methyl 1-(perfluorohex-1-yl)propanoate, Methyl 1-(perfluorohex-1-yl)propionate
PC51089-1G
PC51089-5G
APOSPC51089-1GEA
136
EUR
APOSPC51089-1G
APOSPC51089-5G
Methyl 4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononanoate
Methyl 4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononanoate
Formula:
C₁₀H₇F₁₃O₂ MW: 406,141 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03412256 Núm. CAS: 110260-75-8 |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...