6-(2-Fluoropyridin-3-yl)-9-(tetrahydro-2H-pyran-2-yl)-9H-purine
Proveedor: Apollo Scientific
|
Atención
|
PC51363-1G
PC51363-250MG
APOSPC51363-1GEA
246
EUR
APOSPC51363-1G
APOSPC51363-250MG
6-(2-Fluoropyridin-3-yl)-9-(tetrahydro-2H-pyran-2-yl)-9H-purine
6-(2-Fluoropyridin-3-yl)-9-(tetrahydro-2H-pyran-2-yl)-9H-purine
Can be used to prepare inhibitors of PI3 kinase and/or mTOR and phosphodiesterase 10 inhibitors.
Formula:
C₁₅H₁₄FN₅O Temperatura de almacenaje: Nevera |
Número MDL:
MFCD28402162 Núm. CAS: 1093101-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...