Tris(2,2,2-trifluoroethyl) phosphite 99%
Proveedor: Apollo Scientific
PC52710-10G
PC52710-25G
APOSPC52710-10GEA
256
EUR
APOSPC52710-10G
APOSPC52710-25G
Tris(2,2,2-trifluoroethyl) phosphite 99%
Tris(2,2,2-trifluoroethyl) phosphite
Formula:
C₆H₆F₉O₃P MW: 328,069 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00009902 Núm. CAS: 370-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...