2-(4-Fluorophenoxy)-benzylamine 97%
Proveedor: Apollo Scientific
PC530060-1G
PC530060-500MG
APOSPC530060-1GEA
522
EUR
APOSPC530060-1G
APOSPC530060-500MG
2-(4-Fluorophenoxy)-benzylamine 97%
2-(4-Fluorophenoxy)-benzylamine
Formula:
C₁₃H₁₂FNO MW: 217,239 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD04971088 Núm. CAS: 869945-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...