Methyl 4,5-difluoro-2-iodo-benzoate
Proveedor: Apollo Scientific
PC53611-1G
PC53611-5G
APOSPC53611-1GEA
154
EUR
APOSPC53611-1G
APOSPC53611-5G
Methyl 4,5-difluoro-2-iodo-benzoate
Methyl 4,5-difluoro-2-iodo-benzoate
Formula:
C₈H₅F₂IO₂ Temperatura de almacenaje: Nevera |
Número MDL:
MFCD22490596 Núm. CAS: 1640342-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...