4-Bromo-3-(difluoromethyl)benzoic acid 95%
Proveedor: Apollo Scientific
PC57724-1G
PC57724-250MG
APOSPC57724-1GEA
654
EUR
APOSPC57724-1G
APOSPC57724-250MG
4-Bromo-3-(difluoromethyl)benzoic acid 95%
4-Bromo-3-(difluoromethyl)benzoic acid
Formula:
C₈H₅BrF₂O₂ MW: 251,025 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD32070682 Núm. CAS: 2167386-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...