5-Fluoro-D-tryptophan 97%
Proveedor: Apollo Scientific
PC904537-1G
PC904537-250MG
PC904537-500MG
APOSPC904537-1GEA
870
EUR
APOSPC904537-1G
APOSPC904537-250MG
APOSPC904537-500MG
5-Fluoro-D-tryptophan 97%
5-Fluoro-D-tryptophan
Formula:
C₁₁H₁₁FN₂O₂ MW: 222,216 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06797666 Núm. CAS: 97749-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...