4-Bromo-2-fluorobenzimidamide hydrochloride 97%
Proveedor: Apollo Scientific
PC908612-100MG
PC908612-250MG
APOSPC908612-100MGEA
252
EUR
APOSPC908612-100MG
APOSPC908612-250MG
4-Bromo-2-fluorobenzimidamide hydrochloride 97%
4-Bromo-2-fluorobenzimidamide hydrochloride
Formula:
C₇H₆BrFN₂·ClH MW: 253,499 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD04114441 Núm. CAS: 1187927-25-8 |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...