5-Fluoroisophthalic acid 97%
Proveedor: Apollo Scientific
PC908867-1G
PC908867-5G
APOSPC908867-1GEA
97.3
EUR
APOSPC908867-1G
APOSPC908867-5G
5-Fluoroisophthalic acid 97%
5-Fluoroisophthalic acid
Formula:
C₈H₅FO₄ MW: 184.121 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00230551 Núm. CAS: 1583-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...