Phenethyl caffeate
Proveedor: Apollo Scientific
|
Atención
|
Sinónimos:
3,4-Dihydroxycinnamic Acid Phenethyl Ester, Caffeic Acid Phenethyl Ester, Phenethyl 3-(3,4-dihydroxyphenyl)acrylate, CAPE, Phenethyl 3,4-Dihydroxycinnamate
BIB6022-500MG
BIB6022-1G
BIB6022-5G
APOSBIB6022-500MGEA
36.4
EUR
APOSBIB6022-500MG
APOSBIB6022-1G
APOSBIB6022-5G
Phenethyl caffeate
Phenethyl caffeate
Formula:
C₁₇H₁₆O₄ MW: 284,31 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00866470 Núm. CAS: 104594-70-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...