Memantine lactose adduct 95%

Proveedor: Apollo Scientific

BICL4314-10MG BICL4314-25MG BICL4314-5MG
APOSBICL4314-10MGEA 420 EUR
APOSBICL4314-10MG APOSBICL4314-25MG APOSBICL4314-5MG
Memantine lactose adduct 95%
Memantine lactose adduct
Formula: C₂₄H₄₁NO₁₀
MW: 503,589 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 1159637-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR