(-)-Epigallocatechin gallate

Proveedor: Apollo Scientific
Atención

Sinónimos: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG

BIE4300-50G BIE4300-25G
APOSBIE4300-50GEA 163 EUR
APOSBIE4300-50G APOSBIE4300-25G
(-)-Epigallocatechin gallate
(-)-Epigallocatechin gallate

From green tea.


  • Purity >99% by HPLC/TLC

Formula: C₂₂H₁₈O₁₁
MW: 458,38 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00075940
Núm. CAS: 989-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR