Fusidic acid free acid

Proveedor: Apollo Scientific
Atención

Sinónimos: (Z)-2-((3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetoxy-3,11-dihydroxy-4,8,10,14-tetramethyldodecahydro-1H-cyclopenta[a]phenanthren-17(2H,10H,14H)-ylidene)-6-methylhept-5-enoic acid, Fusidine

BIF6400-1G BIF6400-500MG BIF6400-5G
APOSBIF6400-1GEA 98.6 EUR
APOSBIF6400-1G APOSBIF6400-500MG APOSBIF6400-5G
Fusidic acid free acid
Fusidic acid

Suppresses nitric oxide lysis of pancreatic islet cells.

Formula: C₃₁H₄₈O₆
MW: 516,72 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00865135
Núm. CAS: 6990-06-3
EINECS: 230-256-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR