Motesanib diphosphate
Proveedor: Apollo Scientific
Sinónimos:
N-(3,3-Dimethylindolin-6-yl)-2-((pyridin-4-ylmethyl)amino)nicotinamide diphosphate, N-(3,3-dimethyl-1,2-dihydroindol-6-yl)-2-(pyridin-4-ylmethylamino)pyridine-3-carboxamide phosphoric acid
BIFK0030-10MG
BIFK0030-50MG
BIFK0030-100MG
APOSBIFK0030-10MGEA
65.4
EUR
APOSBIFK0030-10MG
APOSBIFK0030-50MG
APOSBIFK0030-100MG
Motesanib diphosphate
Motesanib diphosphate
Diphosphate Salt, >99%
Formula:
C₂₂H₂₉N₅O₉P₂ MW: 569,4 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD12407403 Núm. CAS: 857876-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...