Methyl-ɑ-D-mannopyranoside

Proveedor: Apollo Scientific
Atención

Sinónimos: ɑ-Methyl-D-mannoside, Methyl-ɑ-D-Mannoside, Methyl α-D-Mannopyranoside, ɑ-Methyl-D-mannopyranoside, (2R,3S,4S,5S,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol

BIM1008-100G BIM1008-25G BIM1008-500G
APOSBIM1008-100GEA 87 EUR
APOSBIM1008-100G APOSBIM1008-25G APOSBIM1008-500G
Methyl-ɑ-D-mannopyranoside
Methyl-alpha-D-mannopyranoside
Formula: C₇H₁₄O₆
MW: 194,18 g/mol
Pto. de fusión: 186…196 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00063262
Núm. CAS: 617-04-9
EINECS: 210-502-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR