2'-Amino-2'-deoxyadenosine
Proveedor: Apollo Scientific
BIMN2004-250MG
BIMN2004-500MG
APOSBIMN2004-250MGEA
406
EUR
APOSBIMN2004-250MG
APOSBIMN2004-500MG
2'-Amino-2'-deoxyadenosine
2'-Amino-2'-deoxyadenosine
Formula:
C₁₀H₁₄N₆O₃ Temperatura de almacenaje: Nevera |
Número MDL:
MFCD06657636 Núm. CAS: 10414-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...