Penicilina G sal sódica Y-IRR
Proveedor: Apollo Scientific
|
Peligro
|
Sinónimos:
6-(2-Phenylacetamido)penicillanic acid sodium salt, Benzylpenicillin sodium salt, Sodium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
BIP0142-100G
BIP0142-250G
BIP0142-25G
APOSBIP0142-100GEA
58
EUR
APOSBIP0142-100G
APOSBIP0142-250G
APOSBIP0142-25G
Penicilina G sal sódica Y-IRR
[2S-(2α,5α,6β)]-6-(Fenilacetamido)-3,3-dimetil-7-oxo-4-tia-1-azabiciclo[3.2.0]heptano-2-carboxilato de sodio
Penicillin G is an inhibitor of bacterial cell wall synthesis.Active against gram-positive and some gram-negative bacteria. b-lactamase sensitive.
- Potency: 1670 mg/mg
Formula:
C₁₆H₁₇N₂NaO₄S Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00069666 Núm. CAS: 69-57-8 EINECS: 200-710-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...