Erastin

Proveedor: Apollo Scientific

Sinónimos: 2-(1-(4-(2-(4-Chlorophenoxy)acetyl)-piperazin-1-yl)-ethyl)-3-(2-ethoxyphenyl)-3H-quinazolin-4-one, 2-(1-(4-(2-(4-Chlorophenoxy)acetyl)piperazin-1-yl)ethyl)-3-(2-ethoxyphenyl)quinazolin-4(3H)-one

BISN0097-1MG BISN0097-5MG
APOSBISN0097-1MGEA 102 EUR
APOSBISN0097-1MG APOSBISN0097-5MG
Erastin
Erastin

Erastin is a compound that interacts with VDAC blocks and reverses mitochondrial depolarization after microtubule destabilizers in intact cells and antagonized tubulin-induced VDAC blockage in planar bilayers It is also a ferroptosis inducer.

Formula: C₃₀H₃₁ClN₄O₄
MW: 547,052 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09837984
Núm. CAS: 571203-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR