(+)-Catechin hydrate

Proveedor: Apollo Scientific

Sinónimos: (+)-Catechin, (+)-3,3',4',5,7-Flavanpentol hydrate, (2R,3S)-2-(3,4-Dihydroxyphenyl)chromane-3,5,7-triol

BISN0111-5MG BISN0111-10MG BISN0111-50MG
APOSBISN0111-5MGEA 87 EUR
APOSBISN0111-5MG APOSBISN0111-10MG APOSBISN0111-50MG
(+)-Catechin hydrate
(+)-Catechin hydrate

(+)-Catechin, a plant secondary metabolite, is an antioxidant flavonoid found in the bark of taxus yunnanensis with anti-atherosclerosis.

Formula: C₁₅H₁₄O₆·xH₂O
MW: 290,27 g/mol (anhy)
Temperatura de almacenaje: Nevera
Número MDL: MFCD00075649
Núm. CAS: 154-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR