IPI-549

Proveedor: Apollo Scientific

BISN0153-1MG BISN0153-5MG BISN0153-10MG
APOSBISN0153-1MGEA 31.9 EUR
APOSBISN0153-1MG APOSBISN0153-5MG APOSBISN0153-10MG
IPI-549
IPI-549

IPI-549 is a selective phosphoinositide-3-kinase (PI3K)-γ inhibitor with IC50 of 16 nM as an immuno-oncology clinical candidate.

Formula: C₃₀H₂₄N₈O₂
Temperatura de almacenaje: Ambiente
Número MDL: MFCD30533720
Núm. CAS: 1693758-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR