1-(4-Methoxyphenyl)-2-(pyrimidin-4-yl)ethanol 95+%
Proveedor: Apollo Scientific
OR0152-1G
APOSOR0152-1GEA
62.5
EUR
APOSOR0152-1G
1-(4-Methoxyphenyl)-2-(pyrimidin-4-yl)ethanol 95+%
1-(4-Methoxyphenyl)-2-(pyrimidin-4-yl)ethanol
Formula:
C₁₃H₁₄N₂O₂ MW: 230,27 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD03426057 Núm. CAS: 185848-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...