1-(4-Azepan-1-ylphenyl)ethanone
Proveedor: Apollo Scientific
OR01991-1G
OR01991-10G
OR01991-5G
APOSOR01991-1GEA
125
EUR
APOSOR01991-1G
APOSOR01991-10G
APOSOR01991-5G
1-(4-Azepan-1-ylphenyl)ethanone
1-(4-Azepan-1-ylphenyl)ethanone
Formula:
C₁₄H₁₉NO MW: 217,31 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD04639769 Núm. CAS: 26586-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...