Ethyl-3-amino-6-bromo-1-benzothiophene-2-carboxylate
Proveedor: Apollo Scientific
OR110207-1G
OR110207-5G
APOSOR110207-1GEA
65.4
EUR
APOSOR110207-1G
APOSOR110207-5G
Ethyl-3-amino-6-bromo-1-benzothiophene-2-carboxylate
Ethyl-3-amino-6-bromo-1-benzothiophene-2-carboxylate
Formula:
C₁₁H₁₀BrNO₂S MW: 300,18 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11505028 Núm. CAS: 1216708-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...