2-(4-Boc-piperazino)-2-(4-methoxyphenyl)acetic acid
Proveedor: Apollo Scientific
OR12722-1G
OR12722-5G
APOSOR12722-1GEA
137
EUR
APOSOR12722-1G
APOSOR12722-5G
2-(4-Boc-piperazino)-2-(4-methoxyphenyl)acetic acid
2-(4-Boc-piperazino)-2-(4-methoxyphenyl)acetic acid
Formula:
C₁₈H₂₆N₂O₅ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD01862497 Núm. CAS: 868260-17-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...