1-Cycloheptenylboronic acid 97%
Proveedor: Apollo Scientific
OR13164-1G
OR13164-250MG
APOSOR13164-1GEA
1030
EUR
APOSOR13164-1G
APOSOR13164-250MG
1-Cycloheptenylboronic acid 97%
1-Cycloheptenylboronic acid
Formula:
C₇H₁₃BO₂ Temperatura de almacenaje: Congelador |
Número MDL:
MFCD04115655 Núm. CAS: 835882-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...