3-(2-Methylphenyl)propionoyl chloride
Proveedor: Apollo Scientific
|
Peligro
|
OR200130-1G
OR200130-250MG
OR200130-500MG
APOSOR200130-1GEA
432
EUR
APOSOR200130-1G
APOSOR200130-250MG
APOSOR200130-500MG
3-(2-Methylphenyl)propionoyl chloride
3-(2-Methylphenyl)propionoyl chloride
Formula:
C₁₀H₁₁ClO Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD16739488 Núm. CAS: 85829-29-4 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...