2-[3-Methyl-2-(methylimino)-4-oxo-1,3-thiazolan-5-yl]acetic acid
Proveedor: Apollo Scientific
OR25955-1G
OR25955-250MG
OR25955-500MG
OR25955-5G
APOSOR25955-1GEA
157
EUR
APOSOR25955-1G
APOSOR25955-250MG
APOSOR25955-500MG
APOSOR25955-5G
2-[3-Methyl-2-(methylimino)-4-oxo-1,3-thiazolan-5-yl]acetic acid
2-[3-Methyl-2-(methylimino)-4-oxo-1,3-thiazolan-5-yl]acetic acid
Formula:
C₇H₁₀N₂O₃S Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD06256825 Núm. CAS: 41306-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...