Benzyl-3-cyano-4-oxopiperidine-1-carboxylate 95+%
Proveedor: Apollo Scientific
OR302400-5G
OR302400-10G
APOSOR302400-5GEA
1030
EUR
APOSOR302400-5G
APOSOR302400-10G
Benzyl-3-cyano-4-oxopiperidine-1-carboxylate 95+%
Benzyl-3-cyano-4-oxopiperidine-1-carboxylate
Formula:
C₁₄H₁₄N₂O₃ MW: 258,28 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09258795 Núm. CAS: 916423-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...