N-(Pyrazinylcarbonyl)-L-phenylalanine
Proveedor: Apollo Scientific
OR302741-1G
OR302741-5G
APOSOR302741-1GEA
0
EUR
APOSOR302741-1G
APOSOR302741-5G
N-(Pyrazinylcarbonyl)-L-phenylalanine
N-(Pyrazinylcarbonyl)-L-phenylalanine
Formula:
C₁₄H₁₃N₃O₃ Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD09800536 Núm. CAS: 114457-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...