Imidazo[1,2-b]pyridazine-6-carboxylic acid 98%
Proveedor: Apollo Scientific
OR308080-1G
OR308080-250MG
OR308080-5G
APOSOR308080-1GEA
741
EUR
APOSOR308080-1G
APOSOR308080-250MG
APOSOR308080-5G
Imidazo[1,2-b]pyridazine-6-carboxylic acid 98%
Imidazo[1,2-b]pyridazine-6-carboxylic acid
Formula:
C₇H₅N₃O₂ MW: 163,1300 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD11044772 Núm. CAS: 769109-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...